missing translation for 'onlineSavingsMsg'
Learn More

Strontium isopropoxide, 96%

Catalog No. AA1448103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1448103 1 g
AA1448106 5 g
AA1448114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1448103 Supplier Thermo Scientific Chemicals Supplier No. 01448103

CAS: 88863-33-6 | C6H14O2Sr | 205.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88863-33-6
Molecular Formula C6H14O2Sr
Molecular Weight (g/mol) 205.80
MDL Number MFCD00050487
InChI Key OHULXNKDWPTSBI-UHFFFAOYSA-N
Synonym strontium isopropoxide, strontium 2+ bis propan-2-olate, strontium propan-2-olate, diisopropoxystrontium, acmc-20egm0, strontium isopropylate, 2-propanol, strontiumsalt 9ci, strontium 2+ ion bis propan-2-olate
PubChem CID 6099237
SMILES [Sr++].CC(C)[O-].CC(C)[O-]

Specifications

Melting Point 130°C (decomposition)
Quantity 1 g
UN Number UN3205
Sensitivity Air and moisture sensitive
Solubility Information Soluble in isoprpanol.
IUPAC Name strontium(2+) bis(propan-2-olate)
Formula Weight 205.82
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Strontium isopropoxide
Hazard Category H228-H252-H314-H335
Hazard Statement GHS H Statement
H314-H318-H228-H252
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Self-heating in large quantities
may catch fire.
Precautionary Statement P210-P235+P410-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: III; Proper Shipping Name: ALKALINE EARTH METAL ALCOHOLATES, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only