missing translation for 'onlineSavingsMsg'
Learn More

Strontium ICP Standard, 1000 ppm Sr in 3% HNO3, Ricca Chemical

Catalog No. MSSR1KN100
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSSR1KN100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSSR1KN100 Supplier Ricca Chemical Company Supplier No. MSSR1KN100

Chemical Identifiers

CAS 10042-76-9 , 7697-37-2 , 7732-18-5
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
PubChem CID 24848
IUPAC Name strontium(2+) dinitrate
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Sr
CAS Min % 94.0, 4.22, 0.23
CAS Max % 97.0, 4.39, 0.24
Equivalence 1 mL = 1 mg Sr
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 100 mL
Grade Laboratory
Chemical Name or Material Strontium ICP Standard
Show More Show Less