missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium ICP Standard, 10,000 ppm Sr in 10% HCl, Ricca Chemical
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| CAS Min % | 85.0, 11.09, 1.44 |
| CAS Max % | 89.0, 11.55, 1.5 |
| Concentration or Composition (by Analyte or Components) | 1mL = 10mg Sr (10, 000ppm Sr) SrCO3 in 10% HCl |
| Chemical Name or Material | Strontium ICP Standard |
Safety and Handling
| ShelfLife | 18 months |