missing translation for 'onlineSavingsMsg'
Learn More

Strontium Bromide, Hexahydrate, Purified, 95%, Spectrum™ Chemical

Catalog No. 18614095
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614095 500 g Poly Bottle
18614094 125 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614095 Supplier Spectrum Chemical Mfg Cor Supplier No. S1665500GM
Only null left

Chemical Identifiers

CAS 7789-53-9

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 95%
Grade Purified
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.