missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strontium AA Standard, 1000 ppm Sr in 3% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural LDPE Bottle |
| Concentration or Composition Notes | 1,000ppm Sr |
| CAS Min % | 94.0, 4.22, 0.23 |
| CAS Max % | 97.0, 4.39, 0.24 |
| Equivalence | 1 mL = 1 mg Sr |
| Chemical Name or Material | Strontium AA Standard |