missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva StrepTrap XT Chromatography Column
StrepTrap XP is a column packed with Strep-Tactin™ XT Sepharose resin to purify Strep-tag™ II and Twin-Strep-tag™ recombinant proteins.
$411.55 - $4268.28
Specifications
| Height (Metric) | 25 mm |
|---|---|
| Column Type | Affinity Column |
| For Use With (Application) | Purification of recombinant proteins tagged with Strep-tag™ II and Twin-Strep-tag™ |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
09920070
|
Cytiva
29401317 |
1 x 1 mL |
Each for $411.55
|
|
|||||
|
09920071
|
Cytiva
29401320 |
5 x 1 mL |
Each for $1,032.33
|
|
|||||
|
09920072
|
Cytiva
29401322 |
1 x 5 mL |
Each for $1,013.50
|
|
|||||
|
09920073
|
Cytiva
29401323 |
5 x 5 mL |
Each for $4,268.28
|
|
|||||
Description
- For use with Strep-Tactin™ XT Sepharose™
- Prepacked in chromatography columns for convenience
Specifications
| 25 mm | |
| Affinity Column | |
| Purification of recombinant proteins tagged with Strep-tag™ II and Twin-Strep-tag™ |