missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva StrepTrap XT Chromatography Column
StrepTrap XP is a column packed with Strep-Tactin™ XT Sepharose resin to purify Strep-tag™ II and Twin-Strep-tag™ recombinant proteins.
Supplier: Cytiva 29401320
Description
- For use with Strep-Tactin™ XT Sepharose™
- Prepacked in chromatography columns for convenience
Spécifications
| 1 mL | |
| 5 x 1 mL | |
| 1 mL/min. | |
| Purification of recombinant proteins tagged with Strep-tag™ II and Twin-Strep-tag™ |
| 7 mm | |
| 25 mm | |
| Affinity Column |