missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Streptavidin, Pacific Orange™ conjugate

Catalog No. 501121545
Encompass
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501121545 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501121545 Supplier Molecular Probes™ Supplier No. S32365

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Streptavidin, Pacific Orange™ conjugate

Pacific Orange™ streptavidin has absorption/emission maxima of ∼400/551 nm. Streptavidin is a nonglycosylated biotin-binding protein that is isolated from bacteria and has a near-neutral isoelectric point. It is widely used to detect biotinylated probes, because it reportedly exhibits less nonspecific binding than does the glycosylated, biotin-binding protein, avidin.

Related Links:

Specifications

Conjugate Pacific Orange
Detection Method Fluorescence
For Use With (Equipment) Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Quantity 1 mg
Content And Storage Contains 1 vial of streptavidin Pacific Orange™ conjugate.

Store in freezer (-5 to -30°C) and protect from light.
Excitation/Emission 400/551
Flow Cytometer Laser Lines 405
Shipping Condition Room Temperature
Form Solid
Product Line Pacific Orange
Product Type Streptavidin Conjugate (Fluorescent)
Label or Dye Pacific Orange
Show More Show Less
I am planning to use a fluorescent streptavidin labeled conjugate. What are the storage conditions and shelf life for the lyophilized powder and reconstituted solution?

In the lyophilized powder form, the fluorescent streptavidin labeled conjugate is stable for six months when stored at -20 degrees C, desiccated, and protected from light. The reconstituted solution is stable for approximately six months when stored at 4 degrees C, protected from light, with the addition of sodium azide to a final concentration of 5 mM or thimerosal to 0.2 mM. For longer storage, we recommend dividing the solution into aliquots and freezing at -20 degrees C, protected from light. Avoid repeated freezing and thawing of the solution.

I am planning to use a fluorescent streptavidin labeled conjugate. How should I prepare the working solution of the conjugate?

The fluorescent streptavidin labeled conjugate solution can be made by dissolving the powder in 0.5-1.0 mL of PBS or other suitable buffer. For details, please refer to page 4 of the "Streptavidin and Fluorescent Conjugates of Streptavidin" manual (https://assets.fishersci.com/TFS-Assets/LSG/manuals/mp00888.pdf).

For Research Use Only. Not for use in diagnostic procedures.