missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin eFluor™ 615 Conjugate

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in immunohistochemistry and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

$424.00 - $610.00

Specifications

Concentration 0.2 mg/mL
Conjugate eFluor 615
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (fluorescent)
View More Specs

Alert:

Contains Sodium azide


Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123673
View Documents
Invitrogen™
42431780
25 μg
Each for $424.00
Only null left
 
501130715
View Documents
Invitrogen™
42431782
100 μg
Each for $610.00
Only null left
 
Description

Description

Specifications

Specifications

0.2 mg/mL
Liquid
Streptavidin Conjugate (fluorescent)
eFluor 615
eBioscience
Biotin
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only