missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin eFluor™ 615 Conjugate

Catalog No. 501123673
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501123673 25 μg
501130715 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Contains Sodium azide
Catalog No. 501123673 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' 42431780
Contains Sodium azide

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in immunohistochemistry and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in immunohistochemistry and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

Emit
615 nm

Excite
595 nm

Reported Application
Immunocytochemistry, Immunohistochemical Staining of Frozen Tissue Sections, Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections

Specifications

Concentration 0.2 mg/mL
Conjugate eFluor 615
Quantity 25 μg
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (fluorescent)
Target Molecule Biotin

For Research Use Only