missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Streptavidin eFluor™ 615 Conjugate

Catalog No. 501130715
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501130715 100 μg
501123673 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Contains Sodium azide
Catalog No. 501130715 Supplier Invitrogen™ Supplier No. 42431782

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains Sodium azide

Commonly used in indirect staining protocols to detect biotinylated primary antibodies in immunohistochemistry and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

The streptavidin fluorochrome conjugates are commonly used in indirect staining protocols to detect biotinylated primary antibodies in immunohistochemistry and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.

Emit
615 nm

Excite
595 nm

Reported Application
Immunocytochemistry, Immunohistochemical Staining of Frozen Tissue Sections, Immunohistochemical Staining of Formalin-Fixed Paraffin Embedded Tissue Sections

Specifications

Concentration 0.2 mg/mL
Conjugate eFluor 615
Quantity 100 μg
Form Liquid
Product Line eBioscience
Product Type Streptavidin Conjugate (fluorescent)
Target Molecule Biotin

For Research Use Only