Learn More
Chemglass Inc Stopcock, Double Oblique, Glass Plug, Pressure, 4mm Bore, 16.2/56 Plug Size, 10mm OD Sidearms
Numéro de catalogue. 50873724
Encompass
Stopcock, Double Oblique, Glass Plug, Pressure, 4mm Bore, 16.2/56 Plug Size, 10mm OD Sidearms. Similar to CG-458, but with two parallel oblique bores. Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug.
Numéro de catalogue. 50873724 Fournisseur Chemglass Inc Code fournisseur CG46002
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Stopcock, Double Oblique, Glass Plug, Pressure, 4mm Bore, 16.2/56 Plug Size, 10mm OD Sidearms. Similar to CG-458, but with two parallel oblique bores. Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug.