Learn More
Chemglass Inc Stopcock, Double Oblique, Hollow Glass Plug, High Vacuum, 4mm Bore, 16.2/56 Plug Size, 10mm OD Sidearms
Numéro de catalogue. CG47402
Encompass
Hollow plug, double oblique bore stopcock is precision ground with corresponding numbers on barrel and plug. Lower vacuum chamber is evacuated with a separate hole in the plug which is not in the line of flow.
Numéro de catalogue. CG47402 Fournisseur Chemglass Inc Code fournisseur CG47402
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Hollow plug, double oblique bore stopcock is precision ground with corresponding numbers on barrel and plug. Lower vacuum chamber is evacuated with a separate hole in the plug which is not in the line of flow.