missing translation for 'onlineSavingsMsg'
Learn More

Labconco Storage Bin Kit for Nexus Horizontal Clean Bench

Catalog No. 17200223
Change view
Click to view available options
For Use With (Equipment):
Enclosures
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17200223 Enclosures
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17200223 Supplier Labconco Supplier No. 5328800
Only null left

Storage Bin kit provides organization for lab supplies and other consumables.

  • Bins mount underneath the work surface of the Nexus Horizontal Clean Bench
  • Includes one bin with mounting hardware
  • Internal bin dimensions: 10 x 7 x 6 in. (25.4 x 17.8 x 15.2 cm)
  • Compatible with Nexus Horizontal Clean Bench only

Specifications

For Use With (Equipment) Enclosures
Product Type Storage Bin Kit