missing translation for 'onlineSavingsMsg'
Learn More

Brady™ STOP Pedestrian Crosswalk Sign

Catalog No. 19129193
Change view
Click to view available options
Material:
Polystyrene
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19129193 Polystyrene
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19129193 Supplier Brady™ Supplier No. 124523
Only null left

Plastic signs for light duty industrial, utility, commercial and institutional environments

  • B-401 plastic signs from Brady™ are ideal for use in light duty industrial, utility, commercial and institutional environments
  • High impact polystyrene with protected graphics
  • Recommended for both indoor and outdoor use
  • Good abrasion and chemical resistance
  • Service temperature of -40°F to 175°F (-40°C to 80°C)
  • Specifications

    Material Polystyrene
    Quantity 1 Ea.
    For Use With (Application) Safety Awareness
    Height (English) 18 in.
    Languages English
    Dimensions (L x W) 18 x 18 in.
    Displayed Text STOP PEDESTRIAN CROSSWALK
    Color White on Red