missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK33 Recombinant Rabbit Monoclonal Antibody (1E4W7)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA538008
Description
Positive Samples: SH-SY5Y Immunogen sequence: KEWKNNPESV EENTTEEKNK PSTEEKLKSY QPWGNVPDAN YTSDEEEEKQ STAYEKQFPA TSKDNFDMCS SSFTSSKLLP AEIKGEMEKT PVTPSQGTAT KYPAKSGALS RTKKKL.
STK33 is a serine/threonine kinase involved in signal transduction and cellular communication. STK33 gene ontology annotations related to this gene include ATP binding; protein kinase activity.Identifiants chimiques
| STK33 | |
| 2.5 mg/mL | |
| ELISA, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q924X7, Q9BYT3 | |
| 117229, 65975 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 399-514 of human STK33 (Q9BYT3). | |
| Antibody |
| 1E4W7 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| STK33 | |
| 4921505G21Rik; AW061264; serine/threonine kinase 33; serine/threonine-protein kinase 33; Stk33 | |
| STK33 | |
| Primary | |
| Affinity chromatography |
Spécifications
| STK33 | |
| Recombinant Monoclonal | |
| 2.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q924X7, Q9BYT3 | |
| STK33 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 399-514 of human STK33 (Q9BYT3). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 1E4W7 | |
| Unconjugated | |
| STK33 | |
| 4921505G21Rik; AW061264; serine/threonine kinase 33; serine/threonine-protein kinase 33; Stk33 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 117229, 65975 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |