missing translation for 'onlineSavingsMsg'
Learn More

STK3 Antibody, ProSci

Numéro de catalogue. 16822130
Modifier l'affichage
Click to view available options
Quantité:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
16822130 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 16822130 Fournisseur ProSci Llc Code fournisseur 429820100

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Goat Polyclonal Antibody

STK3 Polyclonal Antibody specifically detects STK3 in Human, Mouse samples. It is validated for ELISA, Western Blot

Spécifications

Antigène STK3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 500 ug/mL
Conjugué Unconjugated
Dilution Peptide ELISA: antibody detection limit dilution 1:16000.Western Blot:Approx 65kDa band observed in lysates of cell lines A431 and NIH3T3 (calculated MW of 56.3kDa according to Human NP_006272.2 and 56.8kDa according to Mouse NP_062609.2). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. Recommended concentration: 1-3ug/ml.
Formule Supplied at 0.5 mg/mL in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin. Aliquot and store at -20°C. Minimize freezing and thawing.
Numéro d’acquisition du gène NP_006272.2
Alias du gène STK3, serine/threonine kinase 3 (STE20 homolog, yeast), FLJ90748, KRS1, MST2, serine/threonine kinase 3 (Ste20, yeast homolog)
Espèces hôtes Goat
Méthode de purification Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Quantité 0.1 mg
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 6788
Spécifications de test This antibody is not expected to cross-react to STK4
Espèces cibles Human, Mouse
Contenu et stockage Aliquot and store at -20°C. Minimize freezing and thawing.
Type de produit Antibody
Forme Liquid
Isotype IgG
Catégorie de recherche Other
Afficher plus de résultats Afficher moins de résultats