Learn More
Carson™ Stinger™ Series Compact and Lightweight Roof Prism Binoculars
Catalog No. S98180
Encompass
Change view
Click to view available options
Field of View:
288 ft. at 1000 yd.
Catalog No. Field of View
S98180 288 ft. at 1000 yd.
1 options
Catalog No. S98180 Supplier Carson™ Supplier No. HW025
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Stinger Series 10 x 25 mm binoculars feature fully multi-coated lenses and BK-7 prisms housed in a rugged exterior.
  • Lightweight, compact
  • Have fully multi-coated lenses which will allow for bright, sharp and crystal clear images
  • Include a soft pouch, wrist strap and microfiber lens cloth
  • Can be used while traveling, camping, surveillance, nature viewing, bird watching, concerts, sporting events, hunting, safari's and so much more
The Stinger™ binoculars are available in three different sizes:
  • 8 x 22 mm - most compact model and offers the least magnification but the greatest field of view
  • 12 x 32 mm - offer the greatest magnification and best light transmission due to the wider 32 mm objective lenses but the narrowest field of view
  • 10 x 25 mm - falls right in the middle and gives you the best of both worlds
Carson™ only manufactures high quality binoculars and backs their products with a No-Fault, No-Hassle guarantee. In the event that your binoculars get damaged, regardless of cause, Carson™ will repair or replace them at no charge. If your binoculars are found to have manufacturing defects in materials or workmanship, Carson™ will repair or replace your binoculars and return them.
Specifications
Magnification 10x
Lens Coating FMC
Optical Design Roof Prism
Field of View 288 ft. at 1000 yd.