missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STIM1 Monoclonal Antibody (CDN3H4)

Description
This antibody reacts with a cytoplasmic epitope of human and rodent STIM1, a 84 kDa essential and conserved regulator of store-operated Ca2+ channel function. Western Blot: Both reducing and non-reducing conditions; Immunocytochemistry: Methanol-acetone fixation.
STIM1 (stromal interacting molecule; also known as GOK) acts as a sensor of calcium depletion within the endoplasmic reticulum and transduces the signal to Orai1, the presumptive CRAC channel at the plasma membrane. Following decrease of luminal calcium concentration, STIM1 oligomerizes and induces Orai1 to enable entry of extracellular calcium into the cytoplasm. However, the precise mechanism of STIM1-Orai1 interaction has not been elucidated yet. Many questions also remain to be solved around STIM1 functional distribution. It turns out that STIM1 associates with growing ends of microtubules and is involved in endoplasmic reticulum tubule extension.
Spécifications
Spécifications
| Antigen | STIM1 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CDN3H4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | STIM1 |
| Gene Accession No. | P70302, P84903, Q13586 |
| Gene Alias | D11S4896E; GOK; IMD10; RGD:1306831}; SIM; Stim1; stim1 {ECO:0000312; STIM1L; STRMK; Stromal interaction molecule 1; stromal interaction molecule 1 precursor; TAM; TAM1 |
| Afficher plus de résultats |