missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Sterile PETG Media Bottles with HDPE Caps
Gamma-sterilized PETG media bottles feature secure HDPE caps, tamper-evident seals, and leak-proof threading are ideal for safely storing reagents and buffers in lab or biopharma settings with excellent durability and chemical resistance.
$136.11 - $797.71
Specifications
| Color | Clear |
|---|---|
| Material | PETG |
| Sterility | Sterile |
| Closure Included | Yes |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
S139074
|
Eisco Labs
CH5201PK24 |
30 mL |
N/A
|
|
|||||
|
S139057
|
Eisco Labs
CH5202PK24 |
60 mL |
N/A
|
|
|||||
|
S139073
|
Eisco Labs
CH5203PK24 |
125 mL |
N/A
|
|
|||||
|
S139075
|
Eisco Labs
CH5204PK24 |
250 mL |
N/A
|
|
|||||
|
S139076
|
Eisco Labs
CH5205PK24 |
500 mL |
N/A
|
|
|||||
|
S139077
|
Eisco Labs
CH5206PK12 |
1 L |
N/A
|
|
|||||
|
S139078
|
Eisco Labs
CH5207PK6 |
2 L |
N/A
|
|
|||||
Description
- Sterile media bottles - PETG media bottles with secure HDPE caps. Ideal for storing lab reagents, buffers, and media in scientific or biopharmaceutical environments.
- Leak proof and secure - Designed with semi-buttress threads and molded seal rings, ensuring a leak-proof closure for safe transport and storage of sensitive materials.
- Gamma sterilized - Bottles are gamma sterilized per ISO 11137 standards, ensuring safe use for critical laboratory applications.
- Durable and impact-resistant - PETG construction offers exceptional impact resistance, chemical compatibility, and thermal stability from -70°C to 60°C.
- Tamper-evident safety - Each bottle features a tamper-evident heat-shrink seal for added security, ensuring product integrity during storage and use.
Specifications
| Clear | |
| Sterile | |
| Single-use | |
| Narrow | |
| Media Bottle |
| PETG | |
| Yes | |
| No | |
| Cylindrical | |
| Not Autoclavable |