Learn More
Eisco Labs Sterile PETG Media Bottles with HDPE Caps
Gamma-sterilized PETG media bottles feature secure HDPE caps, tamper-evident seals, and leak-proof threading are ideal for safely storing reagents and buffers in lab or biopharma settings with excellent durability and chemical resistance.
Supplier:  Eisco Labs CH5206PK12
 View more versions of this product
Catalog No. S139077
Only null left
Explore available promotions
Description
Description
  • Sterile media bottles - PETG media bottles with secure HDPE caps. Ideal for storing lab reagents, buffers, and media in scientific or biopharmaceutical environments.
  • Leak proof and secure - Designed with semi-buttress threads and molded seal rings, ensuring a leak-proof closure for safe transport and storage of sensitive materials.
  • Gamma sterilized - Bottles are gamma sterilized per ISO 11137 standards, ensuring safe use for critical laboratory applications.
  • Durable and impact-resistant - PETG construction offers exceptional impact resistance, chemical compatibility, and thermal stability from -70°C to 60°C.
  • Tamper-evident safety - Each bottle features a tamper-evident heat-shrink seal for added security, ensuring product integrity during storage and use.
Specifications
Specifications
1 L
Clear
PETG
Sterile
Yes
No
Cylindrical
Media Bottle
33.8 oz.
28.5 mm
12 Pk.
93.1 mm
Single-use
Narrow
220.6 mm
Not Autoclavable
Product Suggestions
Product Suggestions