missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Sterile PETG Media Bottles with HDPE Caps
Gamma-sterilized PETG media bottles feature secure HDPE caps, tamper-evident seals, and leak-proof threading are ideal for safely storing reagents and buffers in lab or biopharma settings with excellent durability and chemical resistance.
Supplier: Eisco Labs CH5204PK24
Description
- Sterile media bottles - PETG media bottles with secure HDPE caps. Ideal for storing lab reagents, buffers, and media in scientific or biopharmaceutical environments.
- Leak proof and secure - Designed with semi-buttress threads and molded seal rings, ensuring a leak-proof closure for safe transport and storage of sensitive materials.
- Gamma sterilized - Bottles are gamma sterilized per ISO 11137 standards, ensuring safe use for critical laboratory applications.
- Durable and impact-resistant - PETG construction offers exceptional impact resistance, chemical compatibility, and thermal stability from -70°C to 60°C.
- Tamper-evident safety - Each bottle features a tamper-evident heat-shrink seal for added security, ensuring product integrity during storage and use.
Specifications
| 250 mL | |
| Clear | |
| PETG | |
| Sterile | |
| Yes | |
| No | |
| Cylindrical | |
| Media Bottle |
| 8.45 oz. | |
| 28.5 mm | |
| 24 Pk. | |
| 61 mm | |
| Single-use | |
| Narrow | |
| 144.6 mm | |
| Not Autoclavable |