Learn More
Eisco Labs Sterile PETG Media Bottles with HDPE Caps
Gamma-sterilized PETG media bottles feature secure HDPE caps, tamper-evident seals, and leak-proof threading are ideal for safely storing reagents and buffers in lab or biopharma settings with excellent durability and chemical resistance.
Supplier:  Eisco Labs CH5205PK24
 View more versions of this product
Catalog No. S139076
Only null left
Explore available promotions
Description
Description
  • Sterile media bottles - PETG media bottles with secure HDPE caps. Ideal for storing lab reagents, buffers, and media in scientific or biopharmaceutical environments.
  • Leak proof and secure - Designed with semi-buttress threads and molded seal rings, ensuring a leak-proof closure for safe transport and storage of sensitive materials.
  • Gamma sterilized - Bottles are gamma sterilized per ISO 11137 standards, ensuring safe use for critical laboratory applications.
  • Durable and impact-resistant - PETG construction offers exceptional impact resistance, chemical compatibility, and thermal stability from -70°C to 60°C.
  • Tamper-evident safety - Each bottle features a tamper-evident heat-shrink seal for added security, ensuring product integrity during storage and use.
Specifications
Specifications
500 mL
Clear
PETG
Sterile
Yes
No
Cylindrical
Media Bottle
17 oz.
28.5 mm
24 Pk.
74.6 mm
Single-use
Narrow
179 mm
Not Autoclavable
Product Suggestions
Product Suggestions