missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Stericup™ E-GP Funnel-less Sterile Vacuum Filtration Systems
GREENER_CHOICE

Catalog No. SEGPU0538
Change view
Click to view available options
Volume (Metric):
500 mL
1000 mL
Outlet Fitting:
38 mm threaded
45 mm threaded
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Outlet Fitting
SEGPU0538 500 mL 38 mm threaded
SEGPU0545 500 mL 45 mm threaded
SEGPU1138 1000 mL 38 mm threaded
SEGPU1145 1000 mL 45 mm threaded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. SEGPU0538 Supplier MilliporeSigma™ Supplier No. SEGPU0538
Get 1 Extra Pack of Filtration Units With Purchase
View all promotions

The newest addition to the Stericup™ Quick Release Family. Evolved with an eco-conscience.

Stericup E and Steritop E sterile vacuum systems are the newest addition to the Stericup Quick Release Family. The Stericup E and Steritop E family of devices are designed as a more sustainable option for sterile filtration that significantly reduces plastic and packaging. These are funnel-less filter products for use in the sterile vacuum filtration of aqueous solutions such as media and tissue culture reagents/buffers.The Stericup E and Steritop E devices are designed specifically for sterile filtration from compatible bottles (38 and 45 mm neck size) of media and tissue culture reagents/buffers. Like Stericup Quick Release filters, Stericup E devices feature a tactile-stop cap to signal secure closure, an ample textured writing surface, and filter membrane characteristics clearly labeled on the filter collar.SEGPU0538 combines a 38 mm threaded collar, which uses a 0.22 μm pore size fast flow PES Express PLUS membrane, with a 500 mL receiver flask for sterilizing tissue culture media and buffers. Not recommended for reagents prepared in other vessel type devices like flasks or anything without a threaded neck.

Specifications

Color Clear with blue adapter
Disposable Single-use
Material Polystyrene
Material (Membrane) PES Express PLUS
Pore Size 0.22 μm
Sterility Sterile
Filtration Area 40 cm²
For Use With (Application) Sterile filtration
Outlet Fitting 38 mm threaded
Outlet Style 38 mm threaded
Type Bottle Top Filter
Volume (Metric) 500 mL
Diameter (Metric) 73 mm
Diameter (Metric) Filter 73 mm
Show More Show Less