Learn More
Stemcell Technologies Inc StemSpan™ Megakaryocyte Expansion Supplement (100X), Size: 1 mL
Numéro de catalogue. NC0890918
Encompass
StemSpan™ Megakaryocyte Expansion Supplement (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of human megakaryocyte progenitor cells from CD34+ cells isolated from human cord blood (CB) or bone marrow (BM) samples.StemSpan™ Megakaryocyte Expansion Supplement (100X) is intended for use in combination with any of the following StemSpan™ media: StemSpan™ SFEM (Catalog #09600) StemSpan™ SFEM II (Catalog #09605) StemSpan™-XF (Catalog #100-0073) StemSpan™-AOF (Catalog #100-0130)When added to serum-free medium, StemSpan™ Megakaryocyte Expansion Supplement typically promotes the production of hundreds of megakaryocytes per input CD34+ cell in 14-day liquid cultures initiated with CD34+ human CB cells. See data tab for more details.
Numéro de catalogue. NC0890918 Fournisseur Stemcell Technologies Inc Code fournisseur 02696
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
StemSpan™ Megakaryocyte Expansion Supplement (100X) contains a combination of recombinant human cytokines formulated to selectively promote the expansion and differentiation of human megakaryocyte progenitor cells from CD34+ cells isolated from human cord blood (CB) or bone marrow (BM) samples.StemSpan™ Megakaryocyte Expansion Supplement (100X) is intended for use in combination with any of the following StemSpan™ media: StemSpan™ SFEM (Catalog #09600) StemSpan™ SFEM II (Catalog #09605) StemSpan™-XF (Catalog #100-0073) StemSpan™-AOF (Catalog #100-0130)When added to serum-free medium, StemSpan™ Megakaryocyte Expansion Supplement typically promotes the production of hundreds of megakaryocytes per input CD34+ cell in 14-day liquid cultures initiated with CD34+ human CB cells. See data tab for more details.