missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ StemPro™ Human Adipose-Derived Stem Cells

Catalog No. R7788115
Encompass
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R7788115 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R7788115 Supplier Gibco™ Supplier No. R7788115

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

StemPro™ Human Adipose-Derived Stem Cells

Human Adipose Derived Stem Cells (ADSCs) are isolated from human lipoaspirate tissue and cryopreserved from primary cultures. Before being cryopreserved the ADSCs are expanded for one passage in MesenPRO RS™ Medium (Cat. no. 12746-012) - a revolutionary new reduced serum (2%) medium that results in reduced ADSC doubling times. Each lot of ADSCs originates from a single donor of human lipoaspirate tissue. Each vial of ADSCs contains cells that can differentiate into multiple mature cell phenotypes in vitro including adipocytes, osteoblasts and chondrocytes. ADSCs have demonstrated very similar phenotypic and functional characteristics to that of bone marrow-derived mesenchymal stem cells. The ADSCs express a flow cytometry cell surface protein profile as follows: positive CD29, CD44, CD73, CD90, CD105, CD166; negative CD14, CD31, CD45, Lin1. For optimal ADSC growth and expansion, MesenPRO RS™ Medium is recommended.

Specifications

Content And Storage StemPro™ Human Adipose Derived Stem Cells (one vial containing ∼1 x 10^6 cells; store in liquid nitrogen)
Cell Type Stem Cell (Mesenchymal)
Form Cryopreserved
Species Human
Tissue Lipoaspirate
Product Line StemPro
Quantity 1 Vial

For Research Use Only. Not for use in diagnostic procedures.