missing translation for 'onlineSavingsMsg'
Learn More

Stearyl Alcohol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012520
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012520 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012520 Supplier Merck Emd Millipore Supplier No. PHR11151G
Only null left
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

Ignition Point 842°F
CAS 112-92-5
Vapor Pressure <0.01 mmHg (38°C)
Vapor Density 9.3 (vs. air)
Density 0.812 g/mL (at 25°C)
Flash Point 195°C
Physical Form Neat
Linear Formula CH3(CH2)17 OH
MDL Number MFCD00002823
Melting Point 56°C to 59°C
Synonym 1-Octadecanol; Octadecyl alcohol; Stearyl alcohol
Type Certified Reference Material
Quantity 1 g
Formula Weight 270.49
Grade Certified Reference Material
Show More Show Less