missing translation for 'onlineSavingsMsg'
Learn More

Stearic Anhydride 97.0+%, TCI America™

Catalog No. S0083500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0083500G 500 g
S008325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0083500G Supplier TCI America Supplier No. S0083500G
Only null left

Chemical Identifiers

CAS 638-08-4
Molecular Formula C36H70O3
Molecular Weight (g/mol) 550.953
MDL Number MFCD00009004
InChI Key WVJVHUWVQNLPCR-UHFFFAOYSA-N
Synonym stearic anhydride, octadecanoic acid, anhydride, stearic acid anhydride, stearicanhydride, octadecanoic acid, 1,1'-anhydride, octadecanoic anhydride, e r degrees eeaou, octadecanoic anhydride #, acmc-1b4gn, octadecanoic acid,1,1'-anhydride
PubChem CID 69485
IUPAC Name octadecanoyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCC

Specifications

Melting Point 71°C
Color White
Quantity 500 g
Formula Weight 550.95
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Stearic Anhydride