Learn More
Stearic anhydride, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 419710250
| Quantity | 25g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C36H70O3 | |
| MFCD00009004 | |
| stearic anhydride, octadecanoic acid, anhydride, stearic acid anhydride, stearicanhydride, octadecanoic acid, 1,1'-anhydride, octadecanoic anhydride, e r degrees eeaou, octadecanoic anhydride #, acmc-1b4gn, octadecanoic acid,1,1'-anhydride | |
| octadecanoyl octadecanoate |
Specifications
| Stearic anhydride | |
| 638-08-4 | |
| Authentic | |
| Glass bottle | |
| (CH3(CH2)16CO)2O | |
| MFCD00009004 | |
| stearic anhydride, octadecanoic acid, anhydride, stearic acid anhydride, stearicanhydride, octadecanoic acid, 1,1'-anhydride, octadecanoic anhydride, e r degrees eeaou, octadecanoic anhydride #, acmc-1b4gn, octadecanoic acid,1,1'-anhydride | |
| CCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCC | |
| 550.95 | |
| 550.95 |
| 98% | |
| 0.8400g/mL | |
| 98% | |
| C36H70O3 | |
| 25g | |
| 0.84 | |
| WVJVHUWVQNLPCR-UHFFFAOYSA-N | |
| octadecanoyl octadecanoate | |
| 69485 | |
| 98% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Warning
EINECSNumber : 211-318-6