missing translation for 'onlineSavingsMsg'
Learn More

Stearic Acid, Powder (Triple Pressed), N.F., J.T. Baker™

Catalog No. 02004346
Change view
Click to view available options
Quantity:
500 g
1.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004346 500 g Poly Bottle
02004322 1.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02004346 Supplier Avantor Supplier No. 034001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57-11-4
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.48
MDL Number MFCD00002752
InChI Key QIQXTHQIDYTFRH-UHFFFAOYSA-N
Synonym stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
PubChem CID 5281
ChEBI CHEBI:28842
IUPAC Name octadecanoic acid
SMILES CCCCCCCCCCCCCCCCCC(O)=O

Specifications

Flash Point 196.00°C
Packaging Poly Bottle
Quantity 500 g
Specific Gravity 1L = 0.94kg
Formula Weight 284.48
Grade NF
Physical Form Powder (Triple Pressed)
Chemical Name or Material Stearic Acid