missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stearic Acid Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 57-11-4 |
| Vapor Pressure | 1 mmHg (173.7°C) |
| Flash Point | 113°C - Closed Cup |
| Physical Form | Neat |
| Linear Formula | CH3(CH2)16 COOH |
| MDL Number | MFCD00002752 |
| Melting Point | 67°C to 72°C |
| Boiling Point | 361°C |
| Synonym | Stearic acid; 1-Heptadecanecarboxylic acid; C18:0; Cetylacetic acid; NSC 25956; NSC 261168; Octadecanoic acid; Stearophanic acid |
| Type | Certified Reference Material |
| Show More |