missing translation for 'onlineSavingsMsg'
Learn More

Stearic acid hydrazide, 97%

Catalog No. AAA1829130
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1829130 250 g
AAA1829109 10 g
AAA1829118 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1829130 Supplier Thermo Scientific Chemicals Supplier No. A1829130
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4130-54-5 | C18H38N2O | 298.515 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4130-54-5
Molecular Formula C18H38N2O
Molecular Weight (g/mol) 298.515
MDL Number MFCD00014764
InChI Key BYTFESSQUGDMQQ-UHFFFAOYSA-N
Synonym stearic acid hydrazide, stearic hydrazide, stearohydrazide, octadecanoic acid, hydrazide, stearoylhydrazine, stearic acid, hydrazide, unii-3fv1c68xop, 3fv1c68xop, octadecanohydrazide #, stearic acid hydrazone
PubChem CID 20088
IUPAC Name octadecanehydrazide
SMILES CCCCCCCCCCCCCCCCCC(=O)NN

Specifications

Melting Point 114°C to 116°C
Quantity 250 g
Formula Weight 298.51
Percent Purity 97%
Chemical Name or Material Stearic acid hydrazide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-946-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only