missing translation for 'onlineSavingsMsg'
Learn More

Stearic Acid, FCC, Spectrum™ Chemical

S1326, 57-11-4

Supplier:  Spectrum Chemical Mfg Cor S132612KG

Quantity 12 kg
Packaging Poly Pail

 View more versions of this product

Catalog No. 18607008


Only null left

Description

Description

Spectrum™ Chemical Stearic Acid, FCC is used as a lubricant, softening and release agent in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

57-11-4 , 57-10-3
284.48
QIQXTHQIDYTFRH-UHFFFAOYSA-N
CCCCCCCCCCCCCCCCCC(O)=O
C18H36O2
MFCD00002752
octadecanoic acid

Specifications

57-11-4 , 57-10-3
360°C
Poly Pail
MFCD00002752
197-212
CCCCCCCCCCCCCCCCCC(O)=O
284.48
Crystalline Powder or Lumps
1
0.001
C18H36O2
12 kg
QIQXTHQIDYTFRH-UHFFFAOYSA-N
octadecanoic acid
FCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

SDS
Documents

Documents

Promotions

Promotions