missing translation for 'onlineSavingsMsg'
Learn More

Stearic Acid, FCC, Spectrum™ Chemical

Catalog No. p-7158315
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607006 500 g Poly Pail
18607007 2.5 kg Poly Pail
18607008 12 kg Poly Pail
18607009 45 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18607006 Supplier Spectrum Chemical Mfg Cor Supplier No. S1326500GM
Only null left

S1326, 57-11-4

Spectrum™ Chemical Stearic Acid, FCC is used as a lubricant, softening and release agent in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 57-11-4 , 57-10-3
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.48
MDL Number MFCD00002752
InChI Key QIQXTHQIDYTFRH-UHFFFAOYSA-N
IUPAC Name octadecanoic acid
SMILES CCCCCCCCCCCCCCCCCC(O)=O

Specifications

CAS Max % 1
Boiling Point 360°C
Ignition Residue 0.001
Packaging Poly Pail
Quantity 500 g
Saponification Value 197-212
Grade FCC
Physical Form Crystalline Powder or Lumps