accessibility menu, dialog, popup

UserName

H3COHO

CAS RN 57-10-3

IUPAC Name: hexadecanoic acid
Synonyms: palmitic acid loxiol EP 278 hydrofol acid 1690 kortacid 1698 palmitinic acid palmitinate prifrac 2960 industrene 4516 hystrene 8016 FAT
Molecular Weight (g/mol): 256.43
Molecular Formula: C16H32O2
InChi Key: IPCSVZSSVZVIGE-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (15)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
Boiling Point 
  • (3)
  • (7)
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (4)
Physical Form 
  • (5)
  • (7)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (7)
  • (2)
  • (7)
  • (3)
  • (7)
  • (4)
  • (15)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (3)
  • (7)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (4)
  • (5)
  • (7)
  • (4)
  • (3)
  • (1)
  • (4)