missing translation for 'onlineSavingsMsg'
Learn More

Stearic Acid, Analytical Standard, ≥98.5%, MilliporeSigma™ Supelco™

Catalog No. 111014688
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014688 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014688 Supplier Merck Emd Millipore Supplier No. 856795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stearic acid is a long chain saturated fatty acid. It can be derived naturally from vegetable and animal fat.

Specifications

CAS 57-11-4
Vapor Pressure 1 mmHg (173.7°C)
Physical Form Neat
Linear Formula CH3(CH2)16 COOH
MDL Number MFCD00002752
Melting Point 67°C to 72°C
Boiling Point 361°C
Synonym 1-Heptadecanecarboxylic acid; C18:0; Cetylacetic acid; NSC 25956; NSC 261168; Octadecanoic acid; Stearophanic acid
Quantity 5 g
Formula Weight 284.48
Percent Purity ≥98.5%
Grade Analytical Standard
Show More Show Less