missing translation for 'onlineSavingsMsg'
Learn More

Stearic acid, ≥98.5%, MilliporeSigma™ Supelco™

Catalog No. 111014689
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014689 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014689 Supplier Merck Emd Millipore Supplier No. 85679500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stearic acid is a long chain saturated fatty acid. It can be derived naturally from vegetable and animal fat.

Specifications

CAS 57-11-4
Vapor Pressure 1 mmHg (173.7°C)
Flash Point 113°C
Shipping Condition Ambient
Physical Form Neat
Linear Formula CH3(CH2)16 COOH
MDL Number MFCD00002752
Melting Point 67°C to 72°C
Boiling Point 361°C
Synonym 1-Heptadecanecarboxylic acid; C18:0; Cetylacetic acid; NSC 25956; NSC 261168; Octadecanoic acid; Stearophanic acid
Type Analytical Standard
Quantity 500 mg
Formula Weight 284.48
Percent Purity ≥98.5%
Grade Analytical Standard
Show More Show Less