missing translation for 'onlineSavingsMsg'
Learn More

Stearic acid, 90+%

Catalog No. AAA176730I
Change view
Click to view available options
Quantity:
1000 g
5000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA176730I 5000 g
AAA176730B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA176730I Supplier Thermo Scientific Chemicals Supplier No. A176730I
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 57-11-4 | C18H36O2 | 284.48 g/mol

Stearic acid is used as a raw material in the production of detergents, surfactants, soaps, and cosmetics such as shampoos and shaving cream products. Its salts are used to soften polyvinyl chloride and a component of grease. It is used for the preparation of softeners in textile sizing with castor oil and dietary supplements. It acts as a hardener in candles with simple sugar or corn syrup. It is utilized for the coating of metal powders like aluminum and iron in fireworks. It finds application as a common lubricant during injection molding and pressing of ceramic powders. It acts as a key raw material for the synthesis of stearyl alcohol by reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Stearic acid is used as a raw material in the production of detergents, surfactants, soaps, and cosmetics such as shampoos and shaving cream products. Its salts are used to soften polyvinyl chloride and a component of grease. It is used for the preparation of softeners in textile sizing with castor oil and dietary supplements. It acts as a hardener in candles with simple sugar or corn syrup. It is utilized for the coating of metal powders like aluminum and iron in fireworks. It finds application as a common lubricant during injection molding and pressing of ceramic powders. It acts as a key raw material for the synthesis of stearyl alcohol by reduction.

Solubility
Soluble in ethanol, dimethyl sulfoxide, dimethyl formamide, carbon terachloride, toluene, chloroform, acetone and carbondisulfide. Insoluble in water.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents and reducing agents.

Chemical Identifiers

CAS 57-11-4
Molecular Formula C18H36O2
Molecular Weight (g/mol) 284.48
MDL Number MFCD00002752
InChI Key QIQXTHQIDYTFRH-UHFFFAOYSA-N
Synonym stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
PubChem CID 5281
ChEBI CHEBI:28842
IUPAC Name octadecanoic acid
SMILES CCCCCCCCCCCCCCCCCC(O)=O

Specifications

Melting Point 67°C to 70°C
Density 0.941
Boiling Point 232°C (15 mmHg)
Flash Point 196°C (384°F)
Odor Odorless
Quantity 5000 g
Beilstein 608585
Merck Index 14,8804
Solubility Information Soluble in ethanol,dimethyl sulfoxide,dimethyl formamide,carbon terachloride,toluene,chloroform,acetone and carbondisulfide. Insoluble in water.
Formula Weight 284.48
Percent Purity ≥90%
Chemical Name or Material Stearic acid
Show More Show Less
Hazard Category H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 200-313-4
RTECSNumber WI2800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only