missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ SteamScrubber Starter Kit

Catalog No. 09311203
Change view
Click to view available options
For Use With (Equipment):
SteamScrubber Glassware Washer
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
09311203 SteamScrubber Glassware Washer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09311203 Supplier Labconco™ Supplier No. 4577100

Outfit a SteamScrubber Glassware Washer to wash a variety of wide-mouth labware.

  • Use to wash a variety of wide-mouth labware
  • Includes Bulk Tube Insert (4573500), Utensil Basket with Cover (4573100), 32-Pin Insert (4401801), and 48-Pin Insert (4574200)

Specifications

Material Stainless Steel
Product Type Starter Kit
Color Silver
No. of Pins 32 or 48
Rack Type Standard
For Use With (Application) Washing Glassware
For Use With (Equipment) SteamScrubber Glassware Washer
Includes Bulk Tube Insert (4573500), Utensil Basket with Cover (4573100), 32-Pin Insert (4401801), and 48-Pin Insert (4574200)
Dimensions (L x W x H) 17.7 x 5.1 x 8.9 in.
Weight (English) Shipping 22 lb.
Weight (Metric) Shipping 9.97 kg
Show More Show Less