missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stationary plug - Medical Nylon - QC

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Nut and body are locked as a single unit. Precision engineered for smooth positive sealing. Color nut for easy recognition. This standard luer is ISO 80369-7 compliant. 

$350.48

Spécifications

Certifications/Compliance Compliance Certificates Available
Description Class 7 Cleanroom Manufactured
For Use With (Application) Tubing connections; Medical device input/output ports; Drug and fluid transfer lines
Packaging Quantity 100 Pack
Storage Requirements Out of direct sunlight, >20°F <100°F
voir les spécifications

Produits 1
Numéro de catalogue Color Prix Quantity & Availability  
Numéro de catalogue Color Prix Quantity & Availability  
01000494
Afficher les documents Détails de la promotion
Natural
paquet de 100 chaque for $350.48
Il en reste null
 
Description

Description

  • Single & Multi Use Applications
  • PVC-Free
  • ISO 80369-7 Compliant
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661, USP81, USP 88
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty

Replacement if not satisfied. 12 Month return Policy.

Spécifications

Spécifications

Compliance Certificates Available
Tubing connections; Medical device input/output ports; Drug and fluid transfer lines
Out of direct sunlight, >20°F <100°F
Class 7 Cleanroom Manufactured
100 Pack
Stationary Luer Lock Plug
Videos
FDS
Documents

Documents