Learn More
ACROBiosystems Inc Recombinant Protein;Human Stathmin 1 / STMN1 Protein;E.coli;100UG;ST1,STMN1,Stathmin,Metablastin,Oncoprotein 18,Op18,C1orf215,LAP18,OP18,pp19,pp17,Protein Pr22
Numéro de catalogue. 501658832
Encompass
Recombinant Protein;Human Stathmin 1 / STMN1 Protein;100UG;Human Stathmin 1, His Tag (ST1-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 149 (Accession # NP_981946).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.1 kDa. The protein migrates as 17-20 kDa under reducing (R) condition (SDS-PAGE).
Numéro de catalogue. 501658832 Fournisseur ACROBiosystems Inc Code fournisseur ST1H5149100UG
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Recombinant Protein;Human Stathmin 1 / STMN1 Protein;100UG;Human Stathmin 1, His Tag (ST1-H5149) is expressed from E.coli cells. It contains AA Ala 2 - Asp 149 (Accession # NP_981946).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 18.1 kDa. The protein migrates as 17-20 kDa under reducing (R) condition (SDS-PAGE).