missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Starter Water Analysis Portable pH Meters

Catalog No. 01922082
Change view
Click to view available options
Model:
ST300
ST300-B
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
01922082 ST300-B
01922081 ST300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01922082 Supplier OHAUS™ Supplier No. 83033962

missing translation for 'validMainframeMsg'

LCD meters combine portability with precise pH measurement.

  • Auto buffer recognition stores calibration data to avoid errors
  • Automatic/manual temperature compensation for accurate readings
  • ORP or temperature electrode connectivity for additional functionality
  • IP54 protection shields water/dust damage
  • Size: 90 x 150 x 35 mm
  • Weight: 0.18 kg

Specifications

Includes 4 AAA Batteries
Accuracy (pH) ±0.01
Data Management Details 30 Sets, most recent calibration data
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±1
Measurement Modes pH with mV and Temperature
Calibration Automatic/Manual
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) -1999 to 1999
Resolution (mV) 1 mV
Calibration Points 1 to 3
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°C
Display Type LCD
Backlight Option None
Data Points 30
Log Function Type Automatic Data Logging
Inputs BNC, NTC
IP Rating IP54
Weight (Metric) 0.18 kg
Material ABS
Model ST300-B
Resolution 0.01 pH; 1 mV; 0.1°C
Battery Type AAA
Type Portable pH Meters
Battery Life >500 hr.
Width (English) 3.5 in.
Width (Metric) 9 cm
Height (English) 1.3 in.
Height (Metric) 3.5 cm
Depth (Metric) 17 cm
Show More Show Less