missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Starter Series Electrode: ST260 2-in-1 pH Electrode
Description
- Designed to produce exact results time and time again
- Sturdy and durable
- Constructed from glass shafts and built to withstand daily use
- Includes a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements
- Fits in electrode holders on bench meters and electrode clips on all portable meters
- Reference System: double salt bridge, Ag/AgCl-KCl
- Salt Bridge Solution: saturated Pure KCl solution
- Reference Electrolyte: 3M KCl Solution
Specifications
- Parameters Measured: pH
- pH Range: 0.00 to 14.00
- Temperature Range: 0° to 100°C
- Connection: BNC
- Cable Length: 1 m
- Shaft dia. x L: 12 x 120 mm
- Zero Potential: 7.00±0.25 pH
- Slope: ≥97%
- Impedance: ≤250MOhm
- Reference Diaphragm: Sleeve
Applications
Ideal for use in lab or in the field.
Specifications
Specifications
| Connector Types | BNC |
| Junction | Ceramic |
| Material | Glass |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0 to 100° |
| Type | 2-in-1 pH Electrode |
| Filling Solution Type | Salt Bridge Solution: saturated Pure KCl solution; Reference Electrolyte: 3M KCl Solution |
| For Use With (Application) | Samples which may react with Ag+ ions such as tri-buffer solution. |
| For Use With (Equipment) | Starter bench and Portable meters |
| Internal Reference | Double salt bridge, Ag/AgCl-KCl |
| Show More |