missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Starter pH Pen Meters

Catalog No. 01922062
Change view
Click to view available options
Display Type:
Dual Line
Single Line
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Type
01922062 Single Line
01922061 Dual Line
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01922062 Supplier OHAUS™ Supplier No. OHS 30073971
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Convenient pH testing. OHAUS Starter pH Pen Meters offer a convenient way to measure pH, or the relative acidity or alkalinity of a sample at a given temperature. The pH pen meters can be completely operated with just one hand, freeing up your other hand to handle samples.

  • Fast, straightforward results
  • Durable IP67 waterproof design
  • Equipped with wrist strap for security
  • Automatic Shutdown after six minutes of non-use
  • Requires four AG13 1.5V Micro Alkaline Batteries
  • Size/Weight: 185 x 45 x 38 mm, 110 g
  • Includes buffer powder pack (ST20)

Specifications

Includes 4 1.5V Batteries
Accuracy (pH) ±0.1
For Use With (Application) Measuring pH, or the relative acidity or alkalinity of a sample at a given temperature
Measurement Modes pH with LogR Temperature
Calibration Automatic/Manual
Range (pH) 0 to 14
Resolution (pH) 0.01
Calibration Points 3
Display Type Single Line
Backlight Option None
Data Points 30
Log Function Type Automatic Data Logging
Weight (Metric) 110 g
Weight (English) 0.24 lb.
Battery Type 4 x AG13 1.5 V Micro Alkaline
Type Portable pH Meter
Show More Show Less