Learn More
Starplex™ Scientific Specimen Containers: Cups
Catalog No. 361031071
Change view
Click to view available options
Capacity (Metric):
90 mL
120 mL
Sterility:
Non-sterile
Closure Type:
Not Included
Wide mouth
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Closure Type
361031071 120 mL Non-sterile Not Included
22146593 90 mL Non-sterile Wide mouth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361031071 Supplier Starplex™ Scientific Supplier No. B1202VO
Only null left
  • Break- and leak-resistant for safety during harsh transport conditions or pneumatic tube systems
  • Clarified polypropylene cups provide easy visual analysis of sample
  • Full-circle thread design prevents cross-threading
  • Graduated markings allow quick measurement of sample
  • Labeling provides ample space for patient information
  • Ridges on cap provide a secure grip for safe handling
Specifications
Closure Included No
Material Polypropylene
Sterility Non-sterile
Capacity (Metric) 120 mL
Closure Type Not Included