missing translation for 'onlineSavingsMsg'
Learn More

Starch, Potato, Reagents

Catalog No. 01336943
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336943 1 kg
01336942 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336943 Supplier Labchem Inc Supplier No. C23315001KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Starch, Potato, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 9005-84-9
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
InChI Key GUBGYTABKSRVRQ-ASMJPISFSA-N
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Boiling Range 668°C
Color White
Boiling Point 668°C
Packaging Poly Bottle
Quantity 1 kg
Specific Gravity 1.5
Grade Reagent
Physical Form Solid
Chemical Name or Material Starch, Potato