missing translation for 'onlineSavingsMsg'
Learn More

Starch-Iodide Solution, Mercury Free, Certified, LabChem™

Catalog No. LC253307
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC253307 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC253307 Supplier LabChem Supplier No. LC253307
Only null left

Specifications

Chemical Name or Material Starch-Iodide Solution
Color Undesignated
Name Note Mercury Free
CAS Max % 99.32, 0.5, 0.13, 0.05
CAS 9005-25-8, 69-72-7, 7681-11-0
Linear Formula KI
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 166
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Suitability Passes Test
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Quantity 125 mL
Physical Form Liquid
Show More Show Less