Learn More
3M™ Petrifilm™ Staph Express Count Plate, Sold by AquaPhoenix Scientific
Catalog No. S30734
Change view
Click to view available options
Quantity:
50 Pack
Catalog No. Quantity
S30734 50 Pack
1 options
Catalog No. S30734 Supplier 3M™ Petrifilm™ Supplier No. CHI6490
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Provides Staphylococcus aureus results
  • Results in 24 hours
  • Plates are sample-ready, eliminating time-consuming steps of preparing media/agar dishes
  • Provides accurate, efficient, consistent results for food processors and contract labs
  • Unique system to facilitates enumeration of S. aureus
  • S. aureus appears as distinctive red-violet colonies
  • This plate has been awarded Official Methods of Analysis (OMA) #2003.07 for processed and prepared foods, #2003.8 for select dairy foods and #2003.11 for Select meat, seafood , poultry and NF VALIDATION certificates: 3M-01/9-04/03 A and 3M-01/9-04/03 B for All human foods and Pet food by AFNOR Certification
Note:
  • Other colonies have to be checked using Staph express disk
Specifications
Detects Staphylococcus aureus
Product Type Count Plate
For Use With (Application) Food processors and contract labs, Facilitating enumeration of S. aureus
Quantity 50 Pack