missing translation for 'onlineSavingsMsg'
Learn More

Stannous Chloride Dihydrate, ACS Reagent (Tin (II) Chloride Dihydrate), Reagents

Catalog No. 01337657
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337657 100 g
01337656 2.5 kg
01337655 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337657 Supplier Labchem Inc Supplier No. C1330000100C2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stannous Chloride Dihydrate, ACS Reagent (Tin (II) Chloride Dihydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10025-69-1
Molecular Formula Cl2H4O2Sn
Molecular Weight (g/mol) 225.64
InChI Key FWPIDFUJEMBDLS-UHFFFAOYSA-L
Synonym Tin(II) Chloride Dihydrate
IUPAC Name λ²-tin(2+) dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Sn++]

Specifications

Boiling Point 652°C
Color White to Off-White
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.71
Chemical Name or Material Stannous Chloride Dihydrate, ACS Reagent (Tin (II) Chloride Dihydrate)