missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Standard Plate Count Agar (APHA) (Dehydrated)

Numéro de catalogue. OXCM0463B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
OXCM0463B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. OXCM0463B Fournisseur Thermo Scientific™ Code fournisseur CM0463B
Il en reste null

Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this Dehydrated Standard Plate Count Agar (APHA)

Determine the plate counts of microorganisms in food, dairy samples, waste water and other materials of sanitary importance with this medium.

  • Meets formulation of APHA, AOAC and PHLS
  • Quality control of Standard Plate Count Agar (APHA) includes testing in accordance with ISO 11133:2014

Spécifications

Certifications/Compliance APHA, AOAC, PHLS, ISO 11133:2014
Form Powder
Product Type Agar
Test Type Standard Plate Count for Microorganism
Quantity 500 g
Yield For 21.3L medium

For laboratory use only